
Citric acid monohydrate ≥98%, crystallised, TECHNICAL
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:5949-29-1
:
Formula:C₆H₈O₇.H₂O
MDL Number:MFCD00149972
:210.14 g/mol
Merck Index:13,02350
Melting Pt:135…152 °C
Density:1.552 g/cm³ (20 °C)
Flash Pt:173.9 °C (closed cup)
EINECS:201-069-1
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Assay:Min. 98 %
Specification Test Results
Related Information
Certificates
Search by Lot Number1L Plastic bottle for solids DIN 60 UN (ZPACK13251000) 33L Plastic bucket UN Z (ZPACK13247037) 5L Plastic bucket UN Z (ZPACK13247005)
Packaging Information
spec_VWRC20278.298 spec_VWRC20278.367 spec_VWRC20278.367 spec_VWRC20278.460 spec_VWRC20278.460
Product specification
VWR Chemicals Big Packs and Bulks
Product Literature
Frequently Bought Together