Order Entry
Czech Republic
ContactUsLinkComponent
Citric acid monohydrate ≥99%, GPR RECTAPUR®
Irritant
Citric acid monohydrate ≥99%, GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates
undefined
Irritant
Citric acid monohydrate ≥99%, GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay:
    Min. 99 %
  • Heavy metals (as Pb):
    Max. 20 ppm
  • Ignition residue (SO4):
    Max. 0.05 %
  • Cl (Chloride):
    Max. 20 ppm
  • SO4 (Sulphate):
    Max. 100 ppm
  • Fe (Iron):
    Max. 20 ppm

Specification Test Results

Frequently Bought Together