Order Entry
Czech Republic
ContactUsLinkComponent
Citric acid monohydrate
Irritant
Citric acid monohydrate
Catalog # HACH1454999
Supplier:  Hach
CAS Number:  
undefined
Irritant
Citric acid monohydrate
Catalog # HACH1454999
Supplier:  Hach
CAS Number:  

Specifications

  • Pk:
    100
  • Seq:
    0100

Specifications

About this item

 :
 :210.14 g/mol
Merck Index:13,02350
Melting Pt:135…152 °C
Density:1.552 g/cm³ (20 °C)
Flash Pt:173.9 °C (closed cup)
EINECS:201-069-1
 :GHS07
Storage Temperature:Ambient
CAS Number:5949-29-1
Formula:C₆H₈O₇.H₂O
MDL Number:MFCD00149972