ContactUsLinkComponent
Silver nitrate 0.1 mol/l (0.1 N) in aqueous solution, AVS TITRINORM Reag. Ph. Eur., USP volumetric solution
Silver nitrate 0.1 mol/l (0.1 N) in aqueous solution, AVS TITRINORM Reag. Ph. Eur., USP volumetric solution
Supplier:  VWR Chemicals
Certificates
undefined
Silver nitrate 0.1 mol/l (0.1 N) in aqueous solution, AVS TITRINORM Reag. Ph. Eur., USP volumetric solution
Supplier:  VWR Chemicals
Certificates
Limited Time Sale
Select items below are currently on sale
Specification Test Results
  • Titer (20°C; real value 0.2 % accuracy):
    0.0998 - 0.1002 mol/l
  • Conforms to Reag. Ph.Eur.:
    Passes test
  • Conforms to USP:
    Passes test
  • NIST traceable:
    Confirmed