Silver nitrate 0.0282 M AgNO3 (1 ml = 1 mg Cl) ≥99.5% volumetric solution, Fluka™
:7761-88-8
:
:AgNO₃
:MFCD00003414
:169.87 g/mol
:Ambient
:231-853-9
- Factor:0.999 - 1.001
- BAM/EMPA reference material:effective
- BAM/EMPA Certificate Number:effective
- Traceable to NIST SRM:effective
Frequently Bought Together