Order Entry
Czech Republic
ContactUsLinkComponent
Sodium thiosulphate 0.0394 M Na2S2O3 (0.0394N) volumetric solution, for the determination of iodine adsorption number (ASTM D 1510), Fluka™
Sodium thiosulphate 0.0394 M Na2S2O3 (0.0394N) volumetric solution, for the determination of iodine adsorption number (ASTM D 1510), Fluka™
Catalog # 35224-1L
CAS Number:  
Sodium thiosulphate 0.0394 M Na2S2O3 (0.0394N) volumetric solution, for the determination of iodine adsorption number (ASTM D 1510), Fluka™
Catalog # 35224-1L
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Factor:
    0.998 - 1.002

Specification Test Results

About this item

 :
 :158.11 g/mol
Boiling Pt:100 °C (1.013 hPa)
Storage Temperature:Ambient
EINECS:231-867-5
CAS Number:7772-98-7
Formula:Na₂S₂O₃
MDL Number:MFCD00003499