Order Entry
Czech Republic
Orders LinkContactUsLinkComponent
 

Sodium thiosulphate standardised solution 0.01 N in aqueous solution

Sodium thiosulphate standardised solution 0.01 N in aqueous solution
Certificates
Sodium thiosulphate standardised solution 0.01 N in aqueous solution
Sodium thiosulphate standardised solution 0.01 N in aqueous solution

2 Options Available Below

Product Details & Documents

CAS No.: 7772-98-7 Molecular Formula: Na2O3S2 Formula Weight: 158.10 Physical Form: Liquid MDL No.: MFCD00003499

Synonyms:  Sodiumthiosulphate, Sodium hyposulphite, Antichlor, di-Sodium thiosulphate
  • CAS Number:7772-98-7
  • Formula:Na₂S₂O₃
  • MDL Number:MFCD00003499
  • Molecular Weight:158.11 g/mol
  • Storage Temperature:Ambient
  • EINECS:231-867-5
Product Family Options

Product Information