Potassium permanganate 0.05 mol/l (0.25 N) in aqueous solution, Titripur® volumetric solution, Supelco®
Catalog # 4.80160.2500
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:2,5 L
- Seq:5096
Specifications
About this item
:
:158.03 g/mol
ADR:9,III
:M-Clarity™ quality level = MQ100
EINECS:231-760-3
UN:3082
:GHS09
Storage Temperature:Ambient
CAS Number:7722-64-7
Formula:KMnO₄
MDL Number:MFCD00011364