Order Entry
Czech Republic
ContactUsLinkComponent
Perchloric acid 1.0 M HClO4 (1.0N) ≥98% volumetric solution, Fluka™
Irritant
Corrosive
Perchloric acid 1.0 M HClO4 (1.0N) ≥98% volumetric solution, Fluka™
Catalog # 34288-1L
CAS Number:  
undefined
Irritant
Corrosive
Perchloric acid 1.0 M HClO4 (1.0N) ≥98% volumetric solution, Fluka™
Catalog # 34288-1L
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Factor:
    0.999 - 1.001

Specification Test Results

About this item

 :
 :100.46 g/mol
Density:1.054 g/cm³
Storage Temperature:Ambient
EINECS:231-512-4
ADR:8,II
 :GHS07
 :GHS05
CAS Number:7601-90-3
Formula:HClO₄
MDL Number:MFCD00011325