
Hydrochloric acid 2 mol/l (2 N), Titripur® volumetric solution, Supelco®
Catalog # 1.09063.1000
Supplier: Merck
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:1 L
- Seq:5037
Specifications
About this item
:
ADR:8,II
:M-Clarity™ quality level = MQ100
:36.46 g/mol
EINECS:231-595-7
UN:1789
:GHS05
Storage Temperature:Ambient
CAS Number:7647-01-0
Formula:HCl
MDL Number:MFCD00011324