
Hydrochloric acid 0.5 M HCl (0.5N) in aqueous solution Reag. Ph. Eur. volumetric solution, Fluka™
:7647-01-0
:
:HCl
:MFCD00011324
:>100 °C (1.013 hPa)
:0 °C
:1.010 g/cm³ (20 °C)
:Ambient
:231-595-7
:1789
:8,III
:36.46 g/mol
:GHS05
- Factor:0.999 - 1.001
- BAM/EMPA reference material:effective
- BAM/EMPA Certificate Number:effective
- Traceable to NIST SRM:effective
Frequently Bought Together