ContactUsLinkComponent
Hydrochloric acid 0.2 mol/l (0.2 N), AVS TITRINORM volumetric solution
undefined
Hydrochloric acid 0.2 mol/l (0.2 N), AVS TITRINORM volumetric solution
  • Titer (20°C; real value 0.2 % accuracy):
    0.1996 - 0.2004 mol/l
  • NIST traceable:
    Confirmed
Frequently Bought Together