ContactUsLinkComponent
Potassium bromate concentrate for dilution to 1/60 mol in aqueous solution, DILUT-IT®, Laboratory Reagent, J.T.Baker®
  4667.
 :  Avantor
 :  
Potassium bromate concentrate for dilution to 1/60 mol in aqueous solution, DILUT-IT®, Laboratory Reagent, J.T.Baker®
  4667.
 :  Avantor
 :  
  • Pk:
    210 mL
  • Seq:
    2545
  • Titer (N):
    0.0998-0.1002
 :
 :167 g/mol
 :Ambient
 :231-829-8
 :1484
 :5.1,II
 :GHS08
 :GHS07
 :7758-01-2
 :KBrO₃
 :MFCD00011359