

Manganistan draselný 0.002 mol/l concentrated aqueous solution, Titrisol®, Supelco®
1.09930.0001
: Merck
:
- Typ balení:Glass ampoule
- Bal.:1 Ampoul
- Sekvence:0501
- Solubility:(20 °C) soluble
- Density:1.002 g/cm3 (20 °C)
- pH:7.3 (H2O, 20 °C)
- Hazard Class:9
- Hazard Class Description:Miscellaneous Hazardous Material
:
:9,III
:M-Clarity™ quality level = MQ100
:231-760-3
:3082
:GHS08
:GHS09
:Ambient
:7722-64-7
:KMnO₄
:MFCD00011364