(±)-1,3-Butanediol for synthesis, Sigma-Aldrich®
Catalog # 8.01964.1000
Supplier: Merck
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:1 L
- Seq:5037
Specifications
Specification Test Results
- Solubility:(20 °C) soluble
- Molar Mass:90.12 g/mol
- Boiling Point:207.5 °C (1.013 hPa)
- Vapor Pressure:0.08 hPa (20 °C)
- Flash Point:121 °C
- Refractive Index:1.4395 (20 °C, 589 nm)
- Explosion Limit:1.8 - 9.4 %(V)
- Density:1.00 g/cm3 (20 °C)
- pH:6 - 7 (1 g/l, H2O)
- Ignition Point:440 °C DIN 51794
Specification Test Results
About this item
:
:90.12 g/mol
Merck Index:13,01566
Boiling Pt:206…208 °C (1013 hPa)
Melting Pt:? -50 °C
Density:1.004…1.006 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
Flash Pt:116 °C
Storage Temperature:Ambient
EINECS:203-529-7
CAS Number:107-88-0
Formula:C₄H₁₀O₂
MDL Number:MFCD00004554