(±)-1,3-Butanediol 99%, extra pure
:107-88-0
:
:C₄H₁₀O₂
:MFCD00004554
:90.12 g/mol
:13,01566
:203…204 °C (760 mmHg)
:–54 °C
:1.01 g/cm³
:109 °C
:Ambient
:203-529-7
- Appearance:Clear liquid
- Infrared spectrum:Conforms
- Refractive index:1.4390 to 1.4410 (20°C, 589 nm)
- Water:≤0.5 %
- GC:≥98.5 %
- Color scale:≤10 APHA
Frequently Bought Together