Order Entry
Czech Republic
Orders LinkContactUsLinkComponent
(±)-2-Octanol, Sigma-Aldrich®

(±)-2-Octanol, Sigma-Aldrich®

Glass bottle, 1 L
Catalog # SIAL74860-1L
Flammable
Irritant
Supplier:   Merck
CAS Number:  123-96-6
Price
1 730,00 Kč
Log in for availability
1 * 1 L
Product Details & Documents
Synonyms:  (±)-2-Hydroxy octane, 2-Capryl alcohol, (±)-Methyl hexyl carbinol, (±)-Hexyl methyl carbinol, DL-2-Octanol, Octan-2-ol
  • CAS Number:123-96-6
  • Formula:C₈H₁₈O
  • MDL Number:MFCD00004591
  • Molecular Weight:130.23 g/mol
  • Merck Index:13,06785
  • Boiling Pt:180 °C (1013 hPa)
  • Melting Pt:–39 °C
  • Density:0.818 g/cm³ (20 °C)
  • Flash Pt:55 °C
  • Storage Temperature:Ambient
  • EINECS:204-667-0
  • UN:1987
  • ADR:3,III
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L

Specifications