Order Entry
Czech Republic
ContactUsLinkComponent
Membrane filters, polyamide membrane, Whatman™
Membrane filters, polyamide membrane, Whatman™
Certificates
undefined
Membrane filters, polyamide membrane, Whatman™

Specifications

  • Catalog No:
  • 514-2551
  • 514-2552
  • WHAT10414006
  • 514-2553
  • 514-2562
  • 514-2563
  • Pore size:
  • 0,45 µm
  • 0,45 µm
  • 0,20 µm
  • 0,45 µm
  • 0,20 µm
  • 0,20 µm
  • Thickness:
  • 110 µm
  • 110 µm
  • 110 µm
  • 110 µm
  • 110 µm
  • 110 µm
  • Water flow rate:
  • 0,45 ml/min/cm² (0,9 bar)
  • 0,45 ml/min/cm² (0,9 bar)
  • 0,2 ml/min/cm² (0,9 bar)
  • 0,45 ml/min/cm² (0,9 bar)
  • 0,2 ml/min/cm² (0,9 bar)
  • 0,2 ml/min/cm² (0,9 bar)
  • Airflow rate:
  • 20 ml/min/cm² (3 mbar)
  • 20 ml/min/cm² (3 mbar)
  • 10 ml/min/cm² (3 mbar)
  • 20 ml/min/cm² (3 mbar)
  • 10 ml/min/cm² (3 mbar)
  • 10 ml/min/cm² (3 mbar)
  • Max. temperature:
  • 135 °C
  • 135 °C
  • 135 °C
  • 135 °C
  • 135 °C
  • 135 °C

Specifications

Frequently Bought Together