

Hydrochloric acid 30%, BAKER INSTRA-ANALYZED®, J.T.Baker®
Catalog # 9600.1000PE
Supplier: Avantor
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 L
- Seq:5037
Specifications
Specification Test Results
- Tungsten (W):max. 0.1
- Yttrium (Y):max. 0.1
- Ytterbium (Yb):max. 0.1
- Zinc (Zn):max. 1
- Zirconium (Zr):max. 0.1
- Assay (w/w):29 - 31 %
- Colour (Hazen):max. 10
- Ammonium (NH4):max. 0.5
- Bromide (Br):max. 10
- Free Chlorine:max. 0.5
- Total Phosphorus:max. 0.01
- Total Sulfur:max. 0.3
- Silver (Ag):max. 1
- Aluminum (Al):max. 1
- Arsenic (As):max. 0.5
- Gold (Au):max. 0.5
- Boron (B):max. 1
- Barium (Ba):max. 0.1
- Beryllium (Be):max. 0.1
- Bismuth (Bi):max. 0.1
- Calcium (Ca):max. 1
- Cadmium (Cd):max. 0.1
- Cerium (Ce):max. 0.1
- Cobalt (Co):max. 0.1
- Chromium (Cr):max. 0.5
- Cesium (Cs):max. 0.1
- Copper (Cu):max. 0.5
- Dysprosium (Dy):max. 0.1
- Erbium (Er):max. 0.1
- Europium (Eu):max. 0.1
- Iron (Fe):max. 1
- Gallium (Ga):max. 0.1
- Gadolinium (Gd):max. 0.1
- Germanium (Ge):max. 1
- Hafnium (Hf):max. 0.1
- Mercury (Hg):max. 0.1
- Holmium (Ho):max. 0.1
- Indium (In):max. 0.1
- Potassium (K):max. 1
- Lanthanum (La):max. 0.1
- Lithium (Li):max. 0.1
- Lutetium (Lu):max. 0.1
- Magnesium (Mg):max. 0.5
- Manganese (Mn):max. 0.1
- Molybdenum (Mo):max. 0.1
- Sodium (Na):max. 1
- Niobium (Nb):max. 0.1
- Neodymium (Nd):max. 0.1
- Nickel (Ni):max. 0.5
- Lead (Pb):max. 0.1
- Palladium (Pd):For Information Only
- Praseodymium (Pr):max. 0.1
- Platinum (Pt):For Information Only
- Rubidium (Rb):max. 0.1
- Rhenium (Re):max. 0.1
- Rhodium (Rh):max. 0.1
- Ruthenium (Ru):max. 0.1
- Antimony (Sb):max. 0.5
- Scandium (Sc):max. 0.1
- Selenium (Se):max. 1
- Samarium (Sm):max. 0.1
- Tin (Sn):max. 0.5
- Strontium (Sr):max. 0.1
- Tantalum (Ta):For Information Only
- Terbium (Tb):max. 0.1
- Tellurium (Te):max. 0.1
- Thorium (Th):max. 0.1
- Titanium (Ti):max. 0.5
- Thallium (Tl):max. 0.1
- Thulium (Tm):max. 0.1
- Uranium (U):max. 0.1
- Vanadium (V):max. 0.5
Specification Test Results
About this item
:
Boiling Pt:110 °C (1013 hPa)
Melting Pt:–30 °C
Density:1.18 g/cm³ (20 °C)
ADR:8,II
:36.46 g/mol
EINECS:231-595-7
UN:1789
:GHS05
:GHS07
Storage Temperature:Ambient
CAS Number:7647-01-0
Formula:HCl
MDL Number:MFCD00011324