Order Entry
Czech Republic
ContactUsLinkComponent
Hydrochloric acid 20%, AnalaR NORMAPUR® analytical reagent
Irritant
Corrosive
Hydrochloric acid 20%, AnalaR NORMAPUR® analytical reagent
Supplier:  VWR Chemicals
Certificates
undefined
Irritant
Corrosive
Hydrochloric acid 20%, AnalaR NORMAPUR® analytical reagent
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay (titr.):
    Min. 20 %
  • Phosphate:
    Max. 0.00005 %
  • Sulfate:
    Max. 0.0001 %
  • Bromide:
    Max. 0.005 %
  • As:
    Max. 0.000001 %
  • Ni:
    Max. 0.000002 %
  • PB:
    Max. 0.000002 %
  • Al:
    Max. 0.000005 %
  • Zn:
    Max. 0.000005 %
  • Fe:
    Max. 0.00002 %
  • Ca:
    Max. 0.00005 %
  • NA:
    Max. 0.00005 %

Specification Test Results

Frequently Bought Together