Bottles, narrow neck, Boston type, round, PPCO, with screw caps, Nalgene®
Supplier: Thermo Fisher Scientific
Certificates
About this item
PPCO, translucent with PP screw cap.
- Excellent chemical resistance
- Heat resistant up to +121 °C, autoclavable with or without contents
- Robust, leakproof
Ideal for storing and shipping liquids.
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904
Specifications
- Catalog No:
- 215-2570
- 215-2564
- 215-2568
- 215-2569
- 215-2573
- 215-2572
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- NAR Certificate of Retirement
- NAR Certificate of Retirement