
Hydrochloric acid 10%, BAKER, Laboratory Reagent
Catalog # 7219.9060
Supplier: Avantor
CAS Number:
Specifications
- Pack type:Plastic container
- Pk:60 L
- Seq:5407
Specifications
Specification Test Results
- Assay:9.5 - 10.5 %
- Appearance:passes test
- Free Chlorine:max. 1 ppm
- Heavy Metals (as Pb):max. 2 ppm
- Identification:passes test
- Residue after Evaporation:max. 0.01 %
- Sulfates (as SO₄):max. 5 ppm
Specification Test Results
About this item
:
ADR:8,II
:36.46 g/mol
EINECS:231-595-7
UN:1789
:GHS07
Storage Temperature:Ambient
CAS Number:7647-01-0
Formula:HCl
MDL Number:MFCD00011324