ContactUsLinkComponent
Formic acid 49 - 51% (by titrimetric analysis) in water, Puriss. p.a. for HPLC, Fluka™
Irritant
Corrosive
Formic acid 49 - 51% (by titrimetric analysis) in water, Puriss. p.a. for HPLC, Fluka™
Certificates
undefined
Irritant
Corrosive
Formic acid 49 - 51% (by titrimetric analysis) in water, Puriss. p.a. for HPLC, Fluka™
Specification Test Results
  • Assay:
    49.0 - 51.0 %
  • Absorbance at 270 nm:
    Max. 0.05
  • Absorbance 280 nm:
    Max. 0.04
  • Absorbance at 290 nm:
    Max. 0.03
  • Absorbance at 300 nm:
    Max. 0.02
  • Absorbance 500 nm:
    Max. 0.02