ContactUsLinkComponent
Tetrahydrofuran-[D₈] 99.5 atom % D for NMR spectroscopy
Flammable
Irritant
Tetrahydrofuran-[D₈] 99.5 atom % D for NMR spectroscopy
Certificates
undefined
Flammable
Irritant
Tetrahydrofuran-[D₈] 99.5 atom % D for NMR spectroscopy
Specification Test Results
  • Appearance:
    Clear colorless Liquid
  • Water:
    ≤0.06 % (Coulometric)
  • D-Enrichment:
    ≥99.4 %
  • Titration with HClO4:
    ≥98.5 % (on dry substance)