Order Entry
Czech Republic
ContactUsLinkComponent
Toluene-[D₈] 100.0 atom % D for NMR spectroscopy
Flammable
Irritant
Health
Toluene-[D₈] 100.0 atom % D for NMR spectroscopy
Catalog # ACRO217440050
CAS Number:  
undefined
Flammable
Irritant
Health
Toluene-[D₈] 100.0 atom % D for NMR spectroscopy
Catalog # ACRO217440050
CAS Number:  

Specifications

  • Pack type:
    Ampoule
  • Pk:
    5 mL
  • Seq:
    2044
  • Pk Info:
    10 x 0.5 ml

Specifications

Specification Test Results

  • Appearance:
    Clear colorless Liquid
  • Water:
    ≤0.01 %
  • D-Enrichment:
    ≥99.95 %

Specification Test Results

About this item

 :
 :100.08 g/mol
Boiling Pt:110 °C (1013 hPa)
Melting Pt:–84 °C
Density:0.94 g/cm³ (20 °C)
Flash Pt:4 °C (closed cup)
Storage Temperature:Refrigerator
EINECS:218-009-5
UN:1294
ADR:3,II
 :GHS02
 :GHS07
 :GHS08
CAS Number:2037-26-5
Formula:C₆D₅CD₃
MDL Number:MFCD00044638