ContactUsLinkComponent
Toluene-[D₈] 99+ atom % D for NMR spectroscopy
Flammable
Irritant
Health
Toluene-[D₈] 99+ atom % D for NMR spectroscopy
Certificates
undefined
Flammable
Irritant
Health
Toluene-[D₈] 99+ atom % D for NMR spectroscopy
Specification Test Results
  • Appearance:
    Clear colorless Liquid
  • Water:
    ≤0.05 %
  • D-Enrichment:
    ≥99.0 %