

Ethylbenzene-[D10] 99+ atom% D for NMR spectroscopy
Catalog # ACRO321360010
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Ampoule
- Pk:1 mL
- Seq:2070
Specifications
Specification Test Results
- Appearance:Clear colorless Liquid
- D-Enrichment:≥99.0 %
Specification Test Results
About this item
:
Boiling Pt:134.5 °C
Density:0.94 g/cm³ (20 °C)
Flash Pt:15 °C
ADR:3,II
:116.09
EINECS:247-292-8
UN:1175
:GHS07
:GHS02
CAS Number:25837-05-2
Formula:D₃CCD₂OC₆D₅