Order Entry
Czech Republic
ContactUsLinkComponent
1,4-Dichlorobenzene-[D4]
1,4-Dichlorobenzene-[D4]
Catalog # EHERC12372100
Supplier:  EHRENSTORFER
CAS Number:  
1,4-Dichlorobenzene-[D4]
Catalog # EHERC12372100
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,1 g
  • Seq:
    3020

Specifications

About this item

 :
 :150.97 g/mol
CAS Number:3855-82-1
Formula:C₆D₄Cl₂