Order Entry
Czech Republic
ContactUsLinkComponent
Methanol-[D₄] 99.8 atom % D + 0.03% (v/v) TMS for NMR spectroscopy
Toxic
Health
Flammable
Methanol-[D₄] 99.8 atom % D + 0.03% (v/v) TMS for NMR spectroscopy
Catalog # ACRO351470075
CAS Number:  
undefined
Toxic
Health
Flammable
Methanol-[D₄] 99.8 atom % D + 0.03% (v/v) TMS for NMR spectroscopy
Catalog # ACRO351470075
CAS Number:  

Specifications

  • Pack type:
    Ampoule
  • Pk:
    7,5 mL
  • Seq:
    2065
  • Pk Info:
    10 x 0.75 ml

Specifications

Specification Test Results

  • Appearance:
    Clear colorless Liquid
  • Water:
    ≤0.05 % (Coulometric)
  • D-Enrichment:
    ≥99.75 %
  • TMS:
    0.020 to 0.040 %

Specification Test Results

About this item

 :
 :36.01 g/mol
Boiling Pt:65.3 °C (1013 hPa)
Melting Pt:–99 °C
Density:0.888 g/cm³ (20 °C)
Flash Pt:12 °C
Storage Temperature:Ambient
EINECS:212-378-6
UN:1230
ADR:3,II
 :GHS06
 :GHS08
 :GHS02
CAS Number:811-98-3
Formula:D₃COD
MDL Number:MFCD00044637