Order Entry
Czech Republic
ContactUsLinkComponent
Methanol-[D₄] 99.8 atom % D + 0.03% (v/v) TMS for NMR spectroscopy
Methanol-[D₄] 99.8 atom % D + 0.03% (v/v) TMS for NMR spectroscopy
Certificates
undefined
Methanol-[D₄] 99.8 atom % D + 0.03% (v/v) TMS for NMR spectroscopy

Specification Test Results

  • Appearance:
    Clear colorless Liquid
  • Water:
    ≤0.05 % (Coulometric)
  • D-Enrichment:
    ≥99.75 %

Specification Test Results

Related Information

Frequently Bought Together