Methanol-[D₄] 99.5 atom % D for NMR spectroscopy
:811-98-3
:
:D₃COD
:MFCD00044637
:36.01 g/mol
:65.3 °C (1013 hPa)
:–99 °C
:0.888 g/cm³ (20 °C)
:12 °C
:Ambient
:212-378-6
:1230
:3,II
:GHS06
:GHS08
:GHS02
- Appearance:Clear colorless Liquid
- Water:≤0.05 % (Coulometric)
- D-Enrichment:≥99.4 %
- HPLC:≥96.0 %
Frequently Bought Together