Chloroform-[D₁] 99.8 atom % D for NMR spectroscopy
ACRO320690075
:
- Pack type:Ampoule
- Pk:7,5 mL
- Seq:2065
- Pk Info:10 x 0.75 ml
- Appearance:Clear colorless Liquid
- Water:≤0.02 % (Coulometric)
- D-Enrichment:≥99.75 %
:
:120.38 g/mol
:60.8 °C (1013 hPa)
:–64 °C
:1.5 g/cm³ (20 °C)
:Refrigerator
:212-742-4
:1888
:6.1,III
:GHS06
:GHS08
:865-49-6
:CDCl₃
:MFCD00000827