

1,1,2,2-Tetrachloroethane-[D₂] 99 atom % D for NMR spectroscopy
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:33685-54-0
:
Formula:Cl₂CDCDCl₂
MDL Number:MFCD00037672
:169.83 g/mol
Boiling Pt:146 °C (1013 hPa)
Melting Pt:–43 °C
Density:1.61 g/cm³
Storage Temperature:Ambient
EINECS:251-634-1
UN:1702
ADR:6.1,II
:GHS06
:GHS09
Specification Test Results
- Appearance:Clear colorless Liquid
- Water:≤0.03 %
- D-Enrichment:≥98.5 %
- NMR (19F-NMR):Consistent with structure
- NMR (31P-NMR):Consistent with structure
- Elemental Analysis:52.4 to 56.8 % C
- Specific optical rotation:+40° to +50° (20°C, 589 nm) (c=1 in methanol)
Specification Test Results
Frequently Bought Together