

Acetone-[D₆] 99.5 atom % D for NMR spectroscopy
Catalog # ACRO320640075
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Ampoule
- Pk:7,5 mL
- Seq:2065
- Pk Info:10 x 0.75 ml
Specifications
Specification Test Results
- Appearance:Clear colorless Liquid
- Water:≤0.05 % (Coulometric)
- D-Enrichment:≥99.4 %
Specification Test Results
About this item
:
:64.03 g/mol
Boiling Pt:55.5 °C (1013 hPa)
Melting Pt:–93 °C
Density:0.87 g/cm³
Flash Pt:–20 °C (closed cup)
Storage Temperature:Ambient
EINECS:211-563-9
UN:1090
ADR:3,II
:GHS07
:GHS02
CAS Number:666-52-4
Formula:D₃CC(O)CD₃
MDL Number:MFCD00044635