ContactUsLinkComponent
Acetone-[D₆] 99+ atom % D for NMR spectroscopy
Irritant
Flammable
Acetone-[D₆] 99+ atom % D for NMR spectroscopy
Certificates
undefined
Irritant
Flammable
Acetone-[D₆] 99+ atom % D for NMR spectroscopy
Specification Test Results
  • Appearance:
    Clear colorless Liquid
  • Water:
    ≤0.1 % (Coulometric)
  • D-Enrichment:
    ≥99.0 %
  • HPLC:
    ≥97.5 %