Order Entry
Czech Republic
ContactUsLinkComponent
Acetonitrile-[D₃] (99% D) for NMR spectroscopy, Sigma-Aldrich®
Flammable
Irritant
Acetonitrile-[D₃] (99% D) for NMR spectroscopy, Sigma-Aldrich®
Catalog # 1.02904.0010
Supplier:  Merck
CAS Number:  
Flammable
Irritant
Acetonitrile-[D₃] (99% D) for NMR spectroscopy, Sigma-Aldrich®
Catalog # 1.02904.0010
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Ampoule, septum-sealed
  • Pk:
    10 mL
  • Seq:
    2275

Specifications

About this item

 :
 :44.03 g/mol
Boiling Pt:80.7 °C (1013 hPa)
Melting Pt:–42 °C
Density:0.844 g/cm³ (20 °C)
Flash Pt:2 °C (closed cup)
Storage Temperature:Ambient
EINECS:218-616-5
UN:1648
ADR:3,II
 :M-Clarity™ quality level = MQ200
 :GHS02
 :GHS07
CAS Number:2206-26-0
Formula:D₃CCN
MDL Number:MFCD00001881