


Xylene (mixture of isomers) 98%, pure
:1330-20-7
:
:C₆H₄(CH₃)₂
:MFCD00077264
:106.17 g/mol
:13,10136
:136…140 °C (760 mmHg)
:–34 °C
:0.865 g/cm³
:23 °C
:Ambient
:215-535-7
:1307
:3,III
:GHS02
:GHS07
:GHS08
- Appearance:Clear colorless Liquid
- Water:≤0.05 %
- GC:≥97 % (isomers and ethylbenzene)
- Residue after evaporation:≤0.01 %
Frequently Bought Together