Order Entry
Czech Republic
ContactUsLinkComponent
 
Glycerine 86 - 88% ≥86-88% (w/w) in aqueous solution for analysis
    
 
undefined
Glycerine 86 - 88% ≥86-88% (w/w) in aqueous solution for analysis
    
 

 

  • Environmentally Preferable:

 

 

  • Appearance:
    Clear colorless Viscous liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4520 to 1.4550 (20°C, 589 nm)
  • Titration Iodometric:
    86.0 to 88.0 %
  • Water:
    12.0 to 14.0 % (K.F.)
  • Sulfated ash:
    ≤0.005 %
  • Density:
    (20/4°C) 1.224 to 1.232 g/ml
  • Substance darkened by H2SO4:
    Passes test
  • Chloride (Cl):
    ≤1 ppm
  • Sulfate (SO4):
    ≤5 ppm
  • Ammonium (NH4):
    ≤5 ppm
  • Arsenic (As):
    ≤0.5 ppm
  • Iron (Fe):
    ≤5 ppm
  • Heavy metals (as Pb):
    ≤1 ppm
  • Free acid (CH3COOH):
    ≤0.002 %
  • Free alkali (NH3):
    ≤0.0005 %
  • Reducing substances:
    ≤0.003 %
  • Fatty acid esters:
    ≤0.05 %

 

Frequently Bought Together