Ultrafiltration membrane filters, Hydrosart®
These membranes are highly hydrophilic and are often preferred for their high protein recovery when processing very dilute solutions.
- Resistance to autoclaving
- Ease of cleaning and extended chemical resistance
- Catalog No:
- SART14459-47-D
- SART14439-47-D
- SART14429-47-D
- SART14439-63-D
- SART14429-63-D
- SART14429-44-D
- SART14459-76-D
- SART14439-76-D
- SART14439-25-D
- SART14459-63-D
- SART14429-25-D
- SART14429-76-D
- SART14459-25-D
- Thickness:
- 180 μm
- 180 μm
- 180 μm
- 180 μm
- 180 μm
- 180 μm
- 180 μm
- 180 μm
- 180 μm
- 180 μm
- 180 μm
- 180 μm
- 180 μm
Frequently Bought Together