ContactUsLinkComponent
Polypropylene microplates, 96- and 384-well
Catalog # 737-0237
Supplier:  EPPENDORF
   Low Manufacturing Impact
undefined
Polypropylene microplates, 96- and 384-well
Catalog # 737-0237
Supplier:  EPPENDORF
   Low Manufacturing Impact
Specifications
  • Packed:
    12×20
  • Sterile:
    -
  • Colour:
    Clear
  • Description:
    Microplates 384/V, white border, PCR clean, LoBind®
  • Seq:
    20
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental certification:
    Renewable Energy Certificate, ISO 14001
  • Working volume:
    5 - 120 µl
  • Pk:
    240
  • Total volume:
    140 µl
Specifications
About this item
These polypropylene microplates feature high transparency and easy to read alphanumeric labelling, making the identification of the correct plate or an individual well quick, easy, and less error-prone. Black and white versions are available for fluorescence or luminescence detection, offering excellent signal-to-noise ratios for improved sensitivity.
  • Highest transparency for better sample visibility
  • OptiTrack® matrix for easy and fast well identification
  • g-safe® for time saving centrifugation up to 6000×g
  • Well designed for maximum sample recovery with raised rim for effective sealing
  • High chemical resistance
All Eppendorf microplates are manufactured in ISO certified clean room conditions. They are available in the purity levels PCR clean (free of human DNA, DNase, RNase and PCR inhibitors) or sterile. Each batch is tested and certified individually.
Microplate Lids
Product Information