Order Entry
Czech Republic
ContactUsLinkComponent
Chromium ≥99.95% (metals basis), sputtering target, Ø 76.2 mm (3.0 in), Thickness: 6.35 mm (0.250 in)
Chromium ≥99.95% (metals basis), sputtering target, Ø 76.2 mm (3.0 in), Thickness: 6.35 mm (0.250 in)
Catalog # 40845.KS
CAS Number:  
Chromium ≥99.95% (metals basis), sputtering target, Ø 76.2 mm (3.0 in), Thickness: 6.35 mm (0.250 in)
Catalog # 40845.KS
CAS Number:  

Specifications

  • Pk:
    1
  • Seq:
    0001

Specifications

About this item

 :
Merck Index:12,02288
Boiling Pt:2469 °C (1013 hPa)
Melting Pt:1840 °C
Density:7.2 g/cm³ (20 °C)
 :52 g/mol
Storage Temperature:Ambient
EINECS:231-157-5
CAS Number:7440-47-3
Formula:Cr
MDL Number:MFCD00010944