ContactUsLinkComponent
Zinc 98+% stable (acc. to UN classification class 4), dust
Environment
Zinc 98+% stable (acc. to UN classification class 4), dust
Certificates
undefined
Environment
Zinc 98+% stable (acc. to UN classification class 4), dust
Certificates
Specification Test Results
  • Appearance:
    Grey Powder
  • Particle Size/Sieve analysis:
    ≤1 % Residue on 150 µm(+100 mesh) sieve
  • Zinc (Zn):
    ≥98.0 %