ContactUsLinkComponent
Cleanroom face masks, Kimtech™ M6, PE/PP/BiCoSof™
Cleanroom face masks, Kimtech™ M6, PE/PP/BiCoSof™
Catalog # 113-0346
Supplier:  KIMBERLY CLARK
undefined
Cleanroom face masks, Kimtech™ M6, PE/PP/BiCoSof™
Catalog # 113-0346
Supplier:  KIMBERLY CLARK
Specifications
  • Type:
    With earloops
  • Colour:
    White
  • Pk:
    500
Specifications
About this item
This gross particulate mask provides basic protection for low-end cleanrooms. It contains no filter.
  • Two-layer design
  • BiCoSof™ fabric inner facing provides comfort
  • Ultrasonic bonding
Suitable for class 1000, ISO 6, M4.5 cleanrooms.