ContactUsLinkComponent
Zinc ≥99.98% (metals basis), foil, Width: 30 cm (12 in), Thickness: 0.25 mm (0.01 in)
Zinc ≥99.98% (metals basis), foil, Width: 30 cm (12 in), Thickness: 0.25 mm (0.01 in)
Certificates
Zinc ≥99.98% (metals basis), foil, Width: 30 cm (12 in), Thickness: 0.25 mm (0.01 in)
Certificates
Specification Test Results
  • Appearance (Color):
    Gray
  • Form:
    Foil
  • Comment:
    0.22 - 0.28 mm thick
  • Width:
    27 - 33 mm
  • Total Metal Impurities:
    0.02% max.