Order Entry
Czech Republic
ContactUsLinkComponent
Copper 99.9% (trace metals basis), shot 1-10 mm
Copper 99.9% (trace metals basis), shot 1-10 mm
Certificates
undefined
Copper 99.9% (trace metals basis), shot 1-10 mm

Specification Test Results

  • Appearance:
    Rust brown Metallic shots
  • Particle size:
    1 to 10 mm
  • Total trace metal impurities:
    ≤0.1 %

Specification Test Results

Related Information

Frequently Bought Together