Praseodymium ≥99.5% (metals basis excluding Ta), rods, Ø 12.7 mm (0.5 in)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7440-10-0
:
Formula:Pr
MDL Number:MFCD00011174
:140.91 g/mol
Storage Temperature:Ambient
EINECS:231-120-3
Frequently Bought Together