Praseodymium 99% (REO, rare earth oxide basis), foil, Thickness: 0.25 mm (0.01 in)
Catalog # 10305.FF
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:1
- Seq:0001
- Pk Info:25×25 mm
Specifications
About this item
:
:140.91 g/mol
Storage Temperature:Ambient
EINECS:231-120-3
CAS Number:7440-10-0
Formula:Pr
MDL Number:MFCD00011174