
Boron ≥99.5% (metals basis), crystalline pieces ≤ 20 mm (0.8 in)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7440-42-8
:
Formula:B
MDL Number:MFCD00134034
:10.81 g/mol
Merck Index:13,01333
Density:1.73 g/cm³ (15 °C)
EINECS:231-151-2
:GHS07
Storage Temperature:Ambient
Frequently Bought Together