
Calcium ≥97% (metals basis), ingot
Catalog # 10128.39
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:700 g
- Seq:3705
Specifications
About this item
:
:40.08 g/mol
Merck Index:13,01644
Boiling Pt:1484 °C (1013 hPa)
Melting Pt:850 °C
Density:1.54 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:213-179-5
UN:1401
ADR:4.3,II
:GHS02
CAS Number:7440-70-2
Formula:Ca
MDL Number:MFCD00085314