Order Entry
Czech Republic
ContactUsLinkComponent
 
Facial tissues, SCOTT®
  KIMB8837
undefined
Facial tissues, SCOTT®
  KIMB8837

 

  • Packed:
    100 sheets/box, 21 boxes per case
  • L×W:
    186×216 mm
  • Seq:
    01
  • Pk:
    2.100

 

 

Premium quality two-ply tissues made from 100% highly refined recycled fibres.

  • Soft, strong and absorbent

Can also be used with the elegant looking chromed dispenser 7820.

Colour: White

 : FSC certified.
 : Supplied in a single sheet dispenser box (carton).